(±)-1-Pentin-3-ol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(±)-Ethinylethylcarbinol, (±)-Ethylethinylcarbinol
B24853.06
B24853.14
B24853.06EA
49.9
EUR
B24853.06
B24853.14
(±)-1-Pentin-3-ol ≥98%
(±)-1-Pentin-3-ol
Purity: 98%
CAS No.: 4187-86-4
Molecular Formula: C5H8O
Formula Weight: 84.12
Flash Point: 29°(84°F)
Refractive Index: 1.4340
Odour: Recognizable
Boiling Point: 123-125°
Beilstein Reference: 1098409
UN No.: 1986
MDL No.: MFCD00004572
Formel:
C₅H₈O Molecular Weight: 84,12 g/mol Siedepunkt: 123…125 °C Dichte: 0,890 Flammpunkt: 29 °C (84 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004572 CAS-Nummer: 4187-86-4 EINECS: 224-063-0 UN: 1986 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...