(±)-3-Chinuclidinol 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: (±)-3-Hydroxy-1-azabicyclo[2.2.2]octan, (±)-3-Hydroxychinuclidin, (±)-Chinuclidin-3-ol

132240050 132240250
ACRO132240050EA 33.3 EUR
ACRO132240050 ACRO132240250
(±)-3-Chinuclidinol 99%
(±)-3-Chinuclidinol
Formel: C₇H₁₃NO
Molecular Weight: 127,19 g/mol
Schmelzpunkt: 217…224 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151326
CAS-Nummer: 1619-34-7
EINECS: 216-578-4
UN: 3259
ADR: 8,II
Merck Index: 12,08266

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 217 to 224 °C
Titration with HClO4 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR