(±)-3-Chinuclidinol

Lieferant: FLUOROCHEM

Synonyme: (±)-3-Hydroxy-1-azabicyclo[2.2.2]octan, (±)-3-Hydroxychinuclidin, (±)-Chinuclidin-3-ol

008851-1G 008851-10G
FLUO008851-1GEA 0 EUR
FLUO008851-1G FLUO008851-10G
(±)-3-Chinuclidinol
(±)-3-Chinuclidinol
Formel: C₇H₁₃NO
Molecular Weight: 127.19 g/mol
Schmelzpunkt: 225…227 °C
MDL: MFCD00151326
CAS-Nummer: 1619-34-7
EINECS: 216-578-4
UN: 3263
ADR: 8 III
Merck Index: 12,08266

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR