(±)-3-Bromcyclohexen 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Bromocyclohexene
335120100
335120500
335122500
ACRO335120100EA
60.7
EUR
ACRO335120100
ACRO335120500
ACRO335122500
(±)-3-Bromcyclohexen 95%
(±)-3-Bromcyclohexen
Formel:
C₆H₉Br Molecular Weight: 161,04 g/mol Siedepunkt: 64…65 °C (15 mmHg) Dichte: 1,38 g/cm³ Flammpunkt: 54 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00013775 CAS-Nummer: 1521-51-3 UN: 1993 ADR: 3,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow to light brown Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5280 to 1.5320 (20°C, 589 nm) |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...