(±)-2-Methylpiperazin ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Methylpiperazine

B24405.14 B24405.22
B24405.14EA 37.7 EUR
B24405.14 B24405.22
(±)-2-Methylpiperazin ≥98%
(±)-2-Methylpiperazin

Purity: 98% CAS No.: 109-07-9 Molecular Formula: C5H12N2 Formula Weight: 100.17 Flash Point: 65°(149°F) Odour: Amine-like Boiling Point: 155-156° Beilstein Reference: 383566 UN No.: 2925 MDL No.: MFCD00005954

Formel: C₅H₁₂N₂
Molecular Weight: 100,16 g/mol
Siedepunkt: 155…156 °C
Schmelzpunkt: 62…66 °C
Flammpunkt: 65 °C (149 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005954
CAS-Nummer: 109-07-9
EINECS: 203-644-2
UN: 2925
ADR: 4.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR