(±)-δ-Decalacton, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (±)-5-Hydroxydecansäurelacton, (±)-δ-Dekalacton, (±)-Decan-5-olid, (±)-Tetrahydro-6-pentyl-2H-pyran-2-on

M-Clarity™ Qualitätsstufe = MQ300

W236128-100G-K W236128-1KG-K W236128-25G-K
SIALW236128-100G-KEA 885 EUR
SIALW236128-100G-K SIALW236128-1KG-K SIALW236128-25G-K
(±)-δ-Decalacton, Sigma-Aldrich®
(±)-δ-Decalacton
Formel: C₁₀H₁₈O₂
Molecular Weight: 170.25 g/mol
MDL: MFCD00006649
CAS-Nummer: 705-86-2
EINECS: 211-889-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR