Bromwasserstoff -Lösung 33% (w/w) 45% (w/v) in Essigsäure
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: Bromwasserstoff, Hydrogenbromid
A14475.0F A14475.AE A14475.AP
A14475.0FEA 414 EUR
A14475.0F A14475.AE A14475.AP
Bromwasserstoff -Lösung 33% (w/w) 45% (w/v) in Essigsäure
Bromwasserstoff
Hydrobromic acid is an indispensable raw material for organic intermediates, inorganic pharmaceuticals, photosensitive materials, dyes and medicines. It is also used in the production of bromine compounds
It is used as an important agent in the industrial processes of isomerization, polymerization, hydration and dehydration and esterification reactions. It plays an essential role for the extraction of mercury from the cinnebar ore.
Formel: HBr
Molecular Weight: 80.91 g/mol
Siedepunkt: -66.7
Schmelzpunkt: -86
Dichte: 1.40
Flammpunkt: 65 °C (149 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011323
CAS-Nummer: 37348-16-6
EINECS: 233-113-0
UN: 3265
ADR: 8 II
Merck Index: 14,4778