Silbernitrat 0.05 mol/l (0.05 N) aqueous solution, Titripur® Volumetric, Supelco®
Lieferant: Merck
Achtung
Synonyme: Höllenstein
MQ100
1.11718.1000 1.11718.1003
1.11718.1000EA 114.75 EUR
1.11718.1000 1.11718.1003
Silbernitrat 0.05 mol/l (0.05 N) aqueous solution, Titripur® Volumetric, Supelco®
Silbernitrat
Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.
Formel: AgNO₃
Molecular Weight: 169.87 g/mol
Siedepunkt: 444
Schmelzpunkt: 210
Lagertemperatur: Raumtemperatur
MDL: MFCD00003414
CAS-Nummer: 7761-88-8
EINECS: 231-853-9
UN: 3082
ADR: 9
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM