Natriumbis(trimethylsilyl)amid 35% in THF
Lieferant: CHEMPUR SILANE
Synonyme:
Natriumbis(trimethylsilyl)amid, HMDS Natriumsalz, N-Natriohexamethyldisilazan, HMDS-Na
SILS09640.25
SILS09640.100
SILNSILS09640.25EA
0
EUR
SILNSILS09640.25
SILNSILS09640.100
Natriumbis(trimethylsilyl)amid 35% in THF
Natriumbis(trimethylsilyl)amid
Formel:
C₆H₁₈NNaSi₂ Molecular Weight: 183.38 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009835 CAS-Nummer: 1070-89-9 EINECS: 213-983-8 UN: 2924 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...