L(+)-Histidin Hydrochlorid Monohydrat ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme: His HCl Monohydrat, (S)-(+)-Histidin Hydrochlorid Monohydrat, (2S)-(+)-Histidin Hydrochlorid Monohydrat, (S)-(+)-2-Amino-3-(4-imidazoyl)propansäure Hydrochlorid Monohydrat, L-3-Imidazol-4-ylalanin Hydrochlorid Monohydrat, H-His-OH.HCl.H2O
A17627.18 A17627.30 A17627.36
A17627.18EA 42.4 EUR
A17627.18 A17627.30 A17627.36
L(+)-Histidin Hydrochlorid Monohydrat ≥99%
L(+)-Histidin Hydrochlorid Monohydrat
Purity: 99% CAS No.: 5934-29-2 Molecular Formula: C6H9N3O2.HCl.H2O Formula Weight: 209.63 (191.62anhy) Melting Point: ca 240° dec. Beilstein Reference: 4168261 Merck Reference: 14,4720 Optical Rotation: +10° (c=5 in 5N HCl) MDL No.: MFCD00151027
Formel: C₆H₉N₃O₂·HCl·H₂O
Molecular Weight: 209.63 g/mol
Schmelzpunkt: ∼ 240 °C (Zers.)
Dichte: 1.485 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00151027
CAS-Nummer: 5934-29-2
EINECS: 211-438-9
Merck Index: 13,04741
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=561d2381f930afe618f78d680790ea9a43f6596c&vid=8aacf6dc4ef5382410cb743498272b48c3c36999&prid=web&clid=SAPDAM