2,2,2-Trifluorethylammoniumchlorid
Lieferant: FLUOROCHEM
Synonyme:
1-Amino-2,2,2-trifluorethan Hydrochlorid, 2,2,2-Trifluorethylamin Hydrochlorid
007517-10G
007517-25G
007517-5G
FLUO007517-10GEA
0
EUR
FLUO007517-10G
FLUO007517-25G
FLUO007517-5G
2,2,2-Trifluorethylammoniumchlorid
2,2,2-Trifluorethylammoniumchlorid
Formel:
C₂H₅ClF₃N Molecular Weight: 135,52 g/mol Schmelzpunkt: 222…226 °C |
MDL:
MFCD00012875 CAS-Nummer: 373-88-6 EINECS: 206-771-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...