2,2,2-Trifluorethylammoniumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Amino-2,2,2-trifluorethan Hydrochlorid, 2,2,2-Trifluorethylamin Hydrochlorid

M-Clarity™ Qualitätsstufe = MQ100

180386-10G 180386-50G
SIAL180386-10GEA 94.9 EUR
SIAL180386-10G SIAL180386-50G
2,2,2-Trifluorethylammoniumchlorid, Sigma-Aldrich®
2,2,2-Trifluorethylammoniumchlorid
Formel: C₂H₅ClF₃N
Molecular Weight: 135,52 g/mol
Schmelzpunkt: 222…226 °C
MDL: MFCD00012875
CAS-Nummer: 373-88-6
EINECS: 206-771-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR