Diethylammoniumchlorid 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Diethylammonium chloride

155890010 155891000 155895000 155890051
ACRO155890010EA 82 EUR
ACRO155890010 ACRO155891000 ACRO155895000 ACRO155890051
Diethylammoniumchlorid 99%
Diethylammoniumchlorid
Formel: C₄H₁₂ClN
Molecular Weight: 109.6 g/mol
Siedepunkt: 320…330 °C (760 mmHg)
Schmelzpunkt: 227…230 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012499
CAS-Nummer: 660-68-4
EINECS: 211-541-9
Merck Index: 13,03138

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 227 to 230 °C
Loss on drying ≤1 %
Titration with HClO4 ≥98.5 %
UV at 420 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR