Dimethylammoniumchlorid 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Dimethylamin Hydrochlorid
155270010
155270050
155275000
155270025
ACRO155270010EA
84.2
EUR
ACRO155270010
ACRO155270050
ACRO155275000
ACRO155270025
Dimethylammoniumchlorid 99%
Dimethylammoniumchlorid
Formel:
C₂H₈ClN Molecular Weight: 81.54 g/mol Schmelzpunkt: 160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012477 CAS-Nummer: 506-59-2 EINECS: 208-046-5 Merck Index: 13,03255 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration Argentometric | ≥98.5 % (on dry substance) |
Water | ≤1 % |
pH | 5 to 6 (5 % in water) |
Sulfate (SO4) | ≤50 ppm |
Iron (Fe) | ≤50 ppm |
Heavy metals (as Pb) | ≤50 ppm |
Insoluble matter | ≤0.005 % (in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...