Dimethylammoniumchlorid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Dimethylamin Hydrochlorid

A12133.0I A12133.0B A12133.30
A12133.0IEA 274 EUR
A12133.0I A12133.0B A12133.30
Dimethylammoniumchlorid ≥98%
Dimethylammoniumchlorid

Purity: 98+% CAS No.: 506-59-2 Molecular Formula: (CH3)2NH.HCl Formula Weight: 81.55 Melting Point: 168-174° Beilstein Reference: 3589311 Merck Reference: 14,3228 MDL No.: MFCD00012477

Formel: C₂H₈ClN
Molecular Weight: 81.54 g/mol
Schmelzpunkt: 168…174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012477
CAS-Nummer: 506-59-2
EINECS: 208-046-5
Merck Index: 13,03255

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR