Dimethylammoniumchlorid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Dimethylamin Hydrochlorid
M-Clarity™ Qualitätsstufe = MQ200
38960-1KG
38960-250G
38960-50G
SIAL38960-1KGEA
109
EUR
SIAL38960-1KG
SIAL38960-250G
SIAL38960-50G
Dimethylammoniumchlorid, Sigma-Aldrich®
Dimethylammoniumchlorid
Formel:
C₂H₈ClN Molecular Weight: 81.54 g/mol Schmelzpunkt: 170…173 °C |
MDL:
MFCD00012477 CAS-Nummer: 506-59-2 EINECS: 208-046-5 Merck Index: 13,03255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...