N,N,N',N'-Tetramethyl-p-phenylendiamin Dihydrochlorid 98+%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: N,N,N',N'-Tetramethyl-p-phenylendiammoniumdichlorid, N,N,N',N'-Tetramethyl-1,4-phenylendiamin Dihydrochlorid, N,N,N',N'-Tetramethyl-1,4-phenylendiammoniumdichlorid, TMPD 2 HCl
420600050 420600250 420601000
ACRO420600050EA 58.4 EUR
ACRO420600050 ACRO420600250 ACRO420601000
N,N,N',N'-Tetramethyl-p-phenylendiamin Dihydrochlorid 98+%
N,N,N',N'-Tetramethyl-p-phenylendiamin Dihydrochlorid
Formel: C₁₀H₁₈Cl₂N₂
Molecular Weight: 237.17 g/mol
Schmelzpunkt: 220…224 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012482
CAS-Nummer: 637-01-4
EINECS: 211-274-8
Merck Index: 12,09367
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e955f6b2a1434ddba239f533c6611583219f8904&vid=13fb110b013db70c026f8eefeee7f08fa2568c0c&prid=web&clid=SAPDAM
Bestellen
Spezifikation Testergebnisse
Appearance White to beige to grey to light brown Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 98.0 to 102.0 %