Bacitracin Zinksalz USP, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
B8800-1250KU
B8800-50KU
SIALB8800-1250KUEA
770
EUR
SIALB8800-1250KU
SIALB8800-50KU
Bacitracin Zinksalz USP, Sigma-Aldrich®
Bacitracin zinc salt
Formel:
C₆₆H₁₀₁N₁₇O₁₆SZn Lagertemperatur: Kühlschrank |
MDL:
MFCD00130598 CAS-Nummer: 1405-89-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...