Acetylene-PEG4-maleimide
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-(N-maleimidyl)-N-(3,6,9,12-tetraoxapentadec-14-yn-1-yl)propionamide
J64859.MB
J64859.MC
J64859.MBEA
155
EUR
J64859.MB
J64859.MC
Acetylene-PEG4-maleimide
Acetylene-PEG4-maleimide
A bifunctional linker with sulfhydryl reactive and azide reactive groups. Contains a PEG spacer to increase solubility