Hydrazin Dihydrochlorid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Hydraziniumdichlorid

32725.22 32725.36
32725.22EA 73.4 EUR
32725.22 32725.36
Hydrazin Dihydrochlorid ≥98%
Hydrazin Dihydrochlorid

Purity: 98+% CAS No.: 5341-61-7 Molecular Formula: H2NNH2.2HCl Formula Weight: 104.97 Physical Form: Lump Density: 1.42 Appearance: White Odour: Odourless Melting Point: 198° Boiling Point: 200° dec. Solubility: Soluble in water. Slightly soluble in alcohol Merck Reference: 14,4770 UN No.: 3288 MDL No.: MFCD00064543


As chlorine scavenger in HCl gas streams

Formel: H₆Cl₂N₂
Molecular Weight: 104,97 g/mol
Siedepunkt: 200 °C (Zersetzung)
Schmelzpunkt: 198 °C
Dichte: 1,42
Lagertemperatur: Raumtemperatur
MDL: MFCD00064543
CAS-Nummer: 5341-61-7
EINECS: 226-283-2
UN: 3288
ADR: 6.1,III
Merck Index: 14,4770

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR