Phenylhydraziniumchlorid 99+%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Hydrazinobenzol Hydrochlorid, Phenylhydrazin Hydrochlorid

151551000 151555000
ACRO151551000EA 66.7 EUR
ACRO151551000 ACRO151555000
Phenylhydraziniumchlorid 99+%
Phenylhydraziniumchlorid
Formel: C₆H₉ClN₂
Molecular Weight: 144,6 g/mol
Schmelzpunkt: 250…254 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012924
CAS-Nummer: 59-88-1
EINECS: 200-444-7
UN: 2811
ADR: 6.1,II
Merck Index: 12,07447
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e16d43c3611e20636f31c95d3974860d83c8913f&vid=c26f549d3aed1b69060ac1042fd1114ca4edb2cb&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow to pink-beige Crystalline powder
Infrared spectrum Conforms
Solubility (5% in H2O) Clear colorless to light yellow solution
HPLC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR