N-(1-Naphthyl)ethylenediamine Dihydrochlorid 96%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: N-(1-Naphthyl)ethylendiamin Dihydrochlorid, N-(1-Naphthyl)ethylendiammoniumdichlorid, Marshall Reagenz, N-2-Aminoethyl-1-naphthylamin Dihydrochlorid, 2-((Aminoethyl)amino)naphthalin Dihydrochlorid
A17164.09 A17164.18
A17164.09EA 54.6 EUR
A17164.09 A17164.18
N-(1-Naphthyl)ethylenediamine Dihydrochlorid 96%
N-2-Aminoethyl-1-naphthylamin Dihydrochlorid
Purity: 96% CAS No.: 1465-25-4 Molecular Formula: C12H14N2.2HCl Formula Weight: 259.18 Melting Point: 191-199° Beilstein Reference: 3707471 Merck Reference: 14,6409 MDL No.: MFCD00012556
Formel: C₁₂H₁₆Cl₂N₂
Molecular Weight: 259.18 g/mol
Schmelzpunkt: 191…199 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012556
CAS-Nummer: 1465-25-4
EINECS: 215-981-2
Merck Index: 12,06496
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3643e6df3e9cf8dc627823e9090c97004ee25308&vid=a30fc28279c11af12c6b735b00b84fc8f08caf34&prid=web&clid=SAPDAM