Zinkacetat Dihydrat ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Zink(II)acetat Dihydrat, Zink(II)acetat-Dihydrat, Zinkdiacetat-Dihydrat, Zinkdiacetat Dihydrat, Essigsäure Zinksalz Dihydrat, Essigsäure Zinksalz-Dihydrat
A12909.0B
A12909.0I
A12909.22
A12909.36
A12909.A4
A12909.0BEA
76.7
EUR
A12909.0B
A12909.0I
A12909.22
A12909.36
A12909.A4
Zinkacetat Dihydrat ≥97%
Zinkacetat Dihydrat
Purity: 97+%
CAS No.: 5970-45-6
Molecular Formula: Zn(OOCCH3)2.2H2O
Formula Weight: 219.50 (183.47anhy)
Odour: Weak, characteristic
pH: 6-7
Melting Point: 237°
Beilstein Reference: 3732513
Merck Reference: 14,10128
UN No.: 3077
MDL No.: MFCD00066961
Formel:
(H₃CCOO)₂Zn·2H₂O Molecular Weight: 219.51 g/mol Schmelzpunkt: 237 Dichte: 1.735 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066961 CAS-Nummer: 5970-45-6 EINECS: 209-170-2 UN: 3077 ADR: 9 III Merck Index: 12,10256 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...