Cobalt(II)acetat Tetrahydrat 98,0-102,0% ACS
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Essigsäure Cobalt(II)salz Tetrahydrat, Cobaltdiacetat Tetrahydrat
44344.14
44344.22
44344.36
44344.14EA
40.8
EUR
44344.14
44344.22
44344.36
Cobalt(II)acetat Tetrahydrat 98,0-102,0% ACS
Cobalt(II)acetat Tetrahydrat
Purity: 98.0-102.0%
CAS No.: 6147-53-1
Molecular Formula: C4H6CoO4.4H2O
Formula Weight: 249.08 (177.02anhy)
Physical Form: Crystalline
Density: 1.71
Refractive Index: 1.542
Sensitivity: Hygroscopic
Appearance: Red
Odour: Odourless
Melting Point: ca 140° -4H2O
Beilstein Reference: 3731397
Merck Reference: 14,2433
UN No.: 3077
MDL No.: MFCD00149569
Formel:
(H₃CCOO)₂Co·4H₂O Molecular Weight: 249,08 g/mol Schmelzpunkt: ∼140 °C (-4H₂O) Dichte: 1,71 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149569 CAS-Nummer: 6147-53-1 EINECS: 200-755-8 UN: 3077 ADR: 9,III Merck Index: 14,2433 |