Nickel(II)acetat Hydrat ≥99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Nickel(II) acetate hydrate

12223.30 12223.A1 12223.A7
12223.30EA 42.7 EUR
12223.30 12223.A1 12223.A7
Nickel(II)acetat Hydrat ≥99%
Nickel(II)acetat Hydrat

Purity: 99+% CAS No.: 6018-89-9 Molecular Formula: Ni(CH3COO)2.xH2O Formula Weight: 176.79(anhy) Physical Form: Crystalline Appearance: Green Odour: Light Melting Point: dec. Solubility: Soluble in water, alcohol Merck Reference: 14,6499 UN No.: 3077 MDL No.: MFCD00012451

Formel: C₄H₆NiO₄·H₂O
Molecular Weight: 194,79 g/mol
Schmelzpunkt: Zersetzung
Lagertemperatur: Raumtemperatur
MDL: MFCD00012451
CAS-Nummer: 6018-89-9
EINECS: 206-761-7
UN: 3077
ADR: 9,III
Merck Index: 14,6499

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR