1-Heptansulfonsäure Natriumsalz Monohydrat ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
Natriumheptylsulfonat Monohydrat
15214.09
15214.18
15214.09EA
73.4
EUR
15214.09
15214.18
1-Heptansulfonsäure Natriumsalz Monohydrat ≥99%
1-Heptansulfonsäure Natriumsalz Monohydrat
Purity: 99%
CAS No.: 207300-90-1
Molecular Formula: C7H15NaO3S.H2O
Formula Weight: 220.26 (202.24anhy)
Physical Form: Crystalline
Appearance: White
Melting Point:
Formel:
C₇H₁₅NaO₃S·1H₂O Molecular Weight: 220.27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149550 CAS-Nummer: 207300-90-1 EINECS: 245-210-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...