Tetrachlorgold(III)säure Hydrat (≥49% Au) ≥99,9% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Tetrachlorgoldsäure Hydrat, Hydrogentetrachloroaurat(III) Hydrat
12325.03
12325.06
12325.14
12325.22
AB202976.0001
AB202976.0005
12325.03EA
189
EUR
12325.03
12325.06
12325.14
12325.22
ABCRAB202976.0001
ABCRAB202976.0005
Tetrachlorgold(III)säure Hydrat (≥49% Au) ≥99,9% (Metall-Basis)
Tetrachlorgold(III)säure Hydrat
Purity: 99.9%
CAS No.: 27988-77-8
Molecular Formula: HAuCl4.xH2O
Formula Weight: 339.79(anhy)
Physical Form: Crystalline
Sensitivity: Light Sensitive and Hygroscopic
Appearance: Yellow-orange
UN No.: 3260
MDL No.: MFCD00149903
Formel:
HAuCl₄·H₂O Molecular Weight: 357,8 g/mol Dichte: 1,169 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149903 CAS-Nummer: 27988-77-8 EINECS: 240-948-4 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...