di-Natriumtetraborat Decahydrat, reinst
Lieferant: Bernd Kraft
|
Gefahr
|
Synonyme:
Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate
06206.3600
06206.5601
06206.2600
06206.4600
KRAF06206.3600EA
54
EUR
KRAF06206.3600
KRAF06206.5601
KRAF06206.2600
KRAF06206.4600
di-Natriumtetraborat Decahydrat, reinst
di-Natriumtetraborat Decahydrat
Formel:
Na₂B₄O₇·10H₂O Molecular Weight: 381.37 g/mol Siedepunkt: 320 Schmelzpunkt: 75 Dichte: 1.73 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149193 CAS-Nummer: 1303-96-4 EINECS: 215-540-4 REACH: 01-2119490790-32 Merck Index: 13,08662 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...