Lithiumchlorid Hydrat ≥99,996% (Metall-Basis), Puratronic®
Lieferant: Thermo Scientific
Achtung
Synonyme: Lithium chloride xhydrate, Hydrochloric acid lithium salt hydrate
10735.14 10735.22
10735.14EA 144 EUR
10735.14 10735.22
Lithiumchlorid Hydrat ≥99,996% (Metall-Basis), Puratronic®
Lithiumchlorid Hydrat
Purity: 99.996% CAS No.: 85144-11-2 Molecular Formula: LiCl.xH2O Formula Weight: 42.39(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 98° -H2O Merck Reference: 14,5528 MDL No.: MFCD00149764
Formel: ClLi·xH₂O
Molecular Weight: 42,39 g/mol (anhy)
Schmelzpunkt: >98 °C (-H₂O)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149764
CAS-Nummer: 85144-11-2
EINECS: 231-212-3
Merck Index: 14,5528