Azadibenzocyclooctyne-amine
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
DBCO-amine, 3-Amino-1-(11,12-didehydrodibenz[b,f]azocin-5(6H)-yl)-1-propanone, Dibenzocyclooctyne-amine
J65637.MB
J65637.MC
J65637.MBEA
178
EUR
J65637.MB
J65637.MC
Azadibenzocyclooctyne-amine
Azadibenzocyclooctyne-amine
A carbonyl reactive reagent used to incorporate ADIBO into organic compounds, surfaces or particles.
Notes: Heat sensitive. Incompatible with amines and azides.
Formel:
C₁₈H₁₆N₂O Molecular Weight: 276,33 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD22380759 CAS-Nummer: 1255942-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...