Azadibenzocyclooctyne-PEG4-maleimide
Lieferant: Thermo Scientific
Achtung
Synonyme: ADIBO-PEG4-maleimide
J65673.MA J65673.MB J65673.MC
J65673.MAEA 125 EUR
J65673.MA J65673.MB J65673.MC
Azadibenzocyclooctyne-PEG4-maleimide
Azadibenzocyclooctyne-PEG4-maleimide
A sulfhydryl reactive reagent with PEG4 spacer for incorporating ADIBO into molecules
Cyclooctynes are useful in strain-promoted copper-free azide-alkyne cycloaddition reactions. This dibenzocyclooctyne will react with azide functionalized compounds or biomolecules without the need for a Cu(I) catalyst to result in a stable triazole linkage. The PEG spacer adds hydrophilicity to the product to decrease aggregation.
Notes: Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.