Azido-PEG3-maleimide Kit

Lieferant: Thermo Scientific
Achtung

Synonyme: N-(11-Azido-3,6,9-trioxaundecyl)-3-(N-maleimidyl)propionamide

J65984.MB J65984.MC
J65984.MBEA 138 EUR
J65984.MB J65984.MC
Azido-PEG3-maleimide Kit
Azido-PEG3-maleimide

An azido containing reagent with PEG spacer and sulfhydryl reactivity. The compound is not stable upon prolonged storage and is sold as a kit.


Notes: Kit contains two components: a) 1.1 eq. of azido-PEG3-amine as an oil; b) 1.0 eq. of maleimide-NHS ester as a solid.

Formel: C₁₅H₂₃N₅O₆
Molecular Weight: 369,37 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD22380764
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR