Bariumperchlorat Trihydrat, Reagenzqualität
Lieferant: Thermo Scientific
Gefahr
11656.30 11656.A1
11656.30EA 65.5 EUR
11656.30 11656.A1
Bariumperchlorat Trihydrat, Reagenzqualität
Bariumperchlorat Trihydrat
CAS No.: 10294-39-0 Molecular Formula: Ba(ClO4)2.3H2O Formula Weight: 390.29 (336.24anhy) Physical Form: Crystalline Density: 2.74 Refractive Index: 1.533 Odour: Odourless Solubility: Soluble in water, methanol. Slightly soluble in ethanol, ethyl acetate, acetone. Insoluble in ether Merck Reference: 14,987 UN No.: 1447 MDL No.: MFCD00149148
Formel: BaCl₂O₈·3H₂O
Molecular Weight: 390,28 g/mol
Schmelzpunkt: 70 °C
Dichte: 2,74
Lagertemperatur: Raumtemperatur
MDL: MFCD00149148
CAS-Nummer: 10294-39-0
EINECS: 236-710-4
UN: 1447
ADR: 5.1,II
Merck Index: 14,987