Bariumiodid Hydrat ≥95%
Lieferant: Thermo Scientific
Achtung
Synonyme: Barium iodide dihydrate
15126.14 15126.22
15126.14EA 75.1 EUR
15126.14 15126.22
Bariumiodid Hydrat ≥95%
Bariumiodid Hydrat
Purity: 95% CAS No.: 13718-50-8 Molecular Formula: BaI2.xH2O Formula Weight: 391.15(anhy) Physical Form: Powder Sensitivity: Light Sensitive and Hygroscopic Appearance: White Odour: Odourless Solubility: Freely soluble in water. Soluble in alcohol and acetone Merck Reference: 14,980 UN No.: 1564 MDL No.: MFCD00003452
Formel: BaI₂·H₂O
Molecular Weight: 409,15 g/mol
Dichte: 5,15 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149155
CAS-Nummer: 7787-33-9
EINECS: 237-276-9
UN: 1564
ADR: 6.1,III
Merck Index: 14,980