Strontiumhydroxid Octahydrat, tech.
Lieferant: Thermo Scientific
|
Gefahr
|
12199.18
12199.A3
12199.A9
12199.18EA
39.1
EUR
12199.18
12199.A3
12199.A9
Strontiumhydroxid Octahydrat, tech.
Strontiumhydroxid Octahydrat
CAS No.: 1311-10-0
Molecular Formula: Sr(OH)2.8H2O
Formula Weight: 265.76 (121.60anhy)
Physical Form: Crystalline
Density: 1.900
Sensitivity: Air Sensitive and Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 100° -8H2O
Merck Reference: 14,8843
UN No.: 3262
MDL No.: MFCD00150395
Formel:
H₂O₂Sr·8H₂O Molecular Weight: 265,76 g/mol Schmelzpunkt: 100 °C (-8H₂O) Dichte: 1,900 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150395 CAS-Nummer: 1311-10-0 EINECS: 242-367-1 UN: 3262 ADR: 8,III Merck Index: 14,8843 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...