Calciumhydrogenphosphat Dihydrat ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Calciumorthophosphat dibasisch Dihydrat, Calciumphosphat dibasisch Dihydrat, Calciumhydrogenorthophosphat Dihydrat

40233.A1 40233.A7
40233.A1EA 128 EUR
40233.A1 40233.A7
Calciumhydrogenphosphat Dihydrat ≥98%
Calciumhydrogenphosphat Dihydrat

Purity: 98% min CAS No.: 7789-77-7 Molecular Formula: CaHO4P.2H2O Formula Weight: 172.09 (136.06anhy) Physical Form: Powder Density: 2.306 Appearance: White Odour: Odourless Melting Point: 109° -H2O Solubility: Slightly soluble in water. Soluble in dilute hydrochloric, nitric, and acetic acid. Insoluble in alcohol Merck Reference: 14,1692 MDL No.: MFCD00149621

Formel: HCaO₄P·2H₂O
Molecular Weight: 172,09 g/mol
Schmelzpunkt: 109 °C (-H₂O)
Dichte: 2,306
Lagertemperatur: Raumtemperatur
MDL: MFCD00149621
CAS-Nummer: 7789-77-7
EINECS: 231-826-1
Merck Index: 14,1692

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR