Capping Reagens B1 40% (v/v) Acetic anhydride in Acetonitril, Novabiochem®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ200
8.57005.0200
8.57005.0500
8.57005.0200EA
0
EUR
8.57005.0200
8.57005.0500
Capping Reagens B1 40% (v/v) Acetic anhydride in Acetonitril, Novabiochem®
Capping Reagens
Dichte:
0.9 g/cm³ Flammpunkt: 9.4 °C |
UN:
2924 ADR: 3 (8) II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...