Ammoniumeisen(II)sulfat Hexahydrat 98,5-101,5% ACS
Lieferant: Thermo Scientific
Achtung
Synonyme: Ferroammoniumsulfat Hexahydrat, Eisen(II)ammoniumsulfat Hexahydrat, Mohrsches Salz
13448.36 13448.A3
13448.36EA 65.3 EUR
13448.36 13448.A3
Ammoniumeisen(II)sulfat Hexahydrat 98,5-101,5% ACS
Ammoniumeisen(II)sulfat Hexahydrat
Purity: 98.5-101.5% CAS No.: 7783-85-9 Molecular Formula: H8FeN2O8S2.6H2O Formula Weight: 392.13 (284.04anhy) Physical Form: Crystalline Density: 1.864 Appearance: Pale bluish green Odour: Odourless Melting Point: ca 100° dec. Solubility: Soluble in water. Insoluble in alcohol Merck Reference: 14,521 MDL No.: MFCD00150530
Polymerization catalyst, photography
Formel: (NH₄)₂Fe(SO₄)₂·6H₂O
Molecular Weight: 392,14 g/mol
Schmelzpunkt: ∼ 100 °C (Zersetzung)
Dichte: 1,864
Lagertemperatur: Raumtemperatur
MDL: MFCD00150530
CAS-Nummer: 7783-85-9
EINECS: 233-382-4
Merck Index: 14,521