Ammoniumeisen(III)sulfat Dodecahydrat 98,5-102,0% ACS
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: Eisen(III)ammoniumsulfat Dodecahydrat, Eisenalaun Dodecahydrat, Eisenammoniumalaun Dodecahydrat, Ammoniumeisenalaun Dodecahydrat
39391.36 39391.A4
39391.36EA 55.5 EUR
39391.36 39391.A4
Ammoniumeisen(III)sulfat Dodecahydrat 98,5-102,0% ACS
Ammoniumeisen(III)sulfat Dodecahydrat
Purity: 98.5-102.0% CAS No.: 7783-83-7 Molecular Formula: H4FeNO8S2.12H2O Formula Weight: 482.18 (266.00anhy) Physical Form: Broken Crystals Density: 1.71 Appearance: Pale purple Odour: Odourless Melting Point: 39-41° Solubility: Freely soluble in water. Insoluble in alcohol Merck Reference: 14,521 MDL No.: MFCD00150004
In dyeing and printing textiles, as analytical lab reagent
Formel: NH₄Fe(SO₄)₂·12H₂O
Molecular Weight: 482.2 g/mol
Siedepunkt: 230
Schmelzpunkt: 39…41 °C
Dichte: 1.71
Lagertemperatur: Raumtemperatur
MDL: MFCD00150004
CAS-Nummer: 7783-83-7
EINECS: 233-382-4
Merck Index: 14,521