Holmiumtrichlorid Hexahydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

289213-25G 289213-5G
SIAL289213-25GEA 242 EUR
SIAL289213-25G SIAL289213-5G
Holmiumtrichlorid Hexahydrat, Sigma-Aldrich®
Holmiumtrichlorid Hexahydrat
Formel: Cl₃Ho·6H₂O
Molecular Weight: 379,38 g/mol
MDL: MFCD00149739
CAS-Nummer: 14914-84-2
EINECS: 233-387-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR