Samarium(III)chlorid, Hyrdrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®

Lieferant: Thermo Scientific
Achtung

Synonyme: Samariumtrichlorid

11231.14 11231.22
11231.14EA 73.1 EUR
11231.14 11231.22
Samarium(III)chlorid, Hyrdrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Samarium(III)chlorid

Purity: 99.9% CAS No.: 10361-82-7 Molecular Formula: SmCl3.xH2O Formula Weight: 256.71(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Merck Reference: 14,8347 MDL No.: MFCD00011235

Formel: Cl₃Sm·xH₂O
Molecular Weight: 256,72 g/mol
Schmelzpunkt: 110 °C (-5H₂O)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011235
CAS-Nummer: 10361-82-7
EINECS: 233-797-0
Merck Index: 14,8347

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR