Terbiumtrinitrat Pentahydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Terbium(III)nitrat Pentahydrat
M-Clarity™ Qualitätsstufe = MQ200
325945-25G
325945-5G
SIAL325945-25GEA
453
EUR
SIAL325945-25G
SIAL325945-5G
Terbiumtrinitrat Pentahydrat, Sigma-Aldrich®
Terbiumtrinitrat Pentahydrat
Formel:
N₃O₉Tb·5H₂O Molecular Weight: 435,02 g/mol Schmelzpunkt: 89,3 °C |
MDL:
MFCD00149867 CAS-Nummer: 57584-27-7 EINECS: 233-138-7 UN: 1477 ADR: 5.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...