Cer(IV)sulfat Tetrahydrat, reinst
Lieferant: Bernd Kraft
|
Achtung
|
Synonyme:
Cerdisulfat Tetrahydrat
15221.0600
15221.2600
15221.1600
KRAF15221.0600EA
0
EUR
KRAF15221.0600
KRAF15221.2600
KRAF15221.1600
Cer(IV)sulfat Tetrahydrat, reinst
Cer(IV)sulfat Tetrahydrat
Formel:
Ce(SO₄)₂·4H₂O Molecular Weight: 404,3 g/mol Schmelzpunkt: 180…200 °C Dichte: 3,91 g/cm³ (20 °C) |
MDL:
MFCD00149427 CAS-Nummer: 10294-42-5 EINECS: 237-029-5 UN: 3260 ADR: 8,III Merck Index: 12,02036 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...