Kupfer(II)nitrat Hydrat ≥99,999% (Metall-Basis), Puratronic®
Lieferant: Thermo Scientific
|
Gefahr
|
10699.06
10699.14
10699.22
10699.36
10699.06EA
27.5
EUR
10699.06
10699.14
10699.22
10699.36
Kupfer(II)nitrat Hydrat ≥99,999% (Metall-Basis), Puratronic®
Kupfer(II)nitrat Hydrat
Purity: 99.999%
CAS No.: 13778-31-9
Molecular Formula: CuN2O6.xH2O
Formula Weight: 187.55(anhy)
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Appearance: Blue
Odour: Odourless
Merck Reference: 14,2643
UN No.: 3085
MDL No.: MFCD00149669
Formel:
CuN₂O₆·H₂O Molecular Weight: 205,57 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00149669 CAS-Nummer: 13778-31-9 EINECS: 221-838-5 UN: 3085 ADR: 5.1,III Merck Index: 14,2643 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...