Yttrium(III)nitrat Tetrahydrat, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Yttriumtrinitrat Tetrahydrat

M-Clarity™ Qualitätsstufe = MQ100

217239-10G 217239-50G
SIAL217239-10GEA 261 EUR
SIAL217239-10G SIAL217239-50G
Yttrium(III)nitrat Tetrahydrat, Sigma-Aldrich®
Yttrium(III)nitrat Tetrahydrat
Formel: N₃O₉Y·4H₂O
Molecular Weight: 346.98 g/mol
MDL: MFCD00149938
CAS-Nummer: 13773-69-8
EINECS: 233-802-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR