Mangan(II)carbonat Hydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Mangancarbonat Hydrat
63539-1KG
63539-250G
SIAL63539-1KGEA
103
EUR
SIAL63539-1KG
SIAL63539-250G
Mangan(II)carbonat Hydrat, Sigma-Aldrich®
Mangan(II)carbonat Hydrat
Formel:
CMnO₃·H₂O Molecular Weight: 132,96 g/mol |
CAS-Nummer:
34156-69-9 EINECS: 209-942-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...