Mangan(II)nitrat Hydrat ≥99,98% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Manganous nitrate hydrate
87848.22
87848.36
87848.22EA
51.5
EUR
87848.22
87848.36
Mangan(II)nitrat Hydrat ≥99,98% (Metall-Basis)
Mangan(II)nitrat Hydrat
Purity: 99.98%
CAS No.: 15710-66-4
Molecular Formula: Mn(NO3)2.xH2O
Formula Weight: 178.95(anhy)
Physical Form: Lump
Sensitivity: Hygroscopic
Appearance: Pink
Odour: Odourless
Melting Point: ca 26°
Merck Reference: 14,5733
UN No.: 2724
MDL No.: MFCD00149788
Formel:
Mn(NO₃)₂·H₂O Molecular Weight: 196,96 g/mol Schmelzpunkt: ∼ 26 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149788 CAS-Nummer: 15710-66-4 EINECS: 233-828-8 UN: 2724 ADR: 5.1,III Merck Index: 14,5733 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...