Nickel(II)bromid Trihydrat ≥98%
Lieferant: Thermo Scientific
Gefahr
40242.18 40242.30
40242.18EA 120 EUR
40242.18 40242.30
Nickel(II)bromid Trihydrat ≥98%
Nickel(II)bromid Trihydrat
Purity: 98% CAS No.: 7789-49-3 Molecular Formula: NiBr2.3H2O Formula Weight: 272.58 (218.53anhy) Physical Form: Crystalline Granular Sensitivity: Hygroscopic Solubility: Soluble in water, alcohol Merck Reference: 14,6502 UN No.: 3077 MDL No.: MFCD00011141
Formel: Br₂Ni·3H₂O
Molecular Weight: 272,54 g/mol
Schmelzpunkt: 300 °C (-3H₂O)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011141
CAS-Nummer: 7789-49-3
EINECS: 236-665-0
UN: 3077
ADR: 9,III
Merck Index: 14,6502