Cobalt(II)chlorid Hexahydrat 98,0-102,0% ACS
Lieferant: Thermo Scientific
Gefahr
Synonyme: Cobaltdichlorid Hexahydrat
36554.22 36554.36
36554.22EA 69.6 EUR
36554.22 36554.36
Cobalt(II)chlorid Hexahydrat 98,0-102,0% ACS
Cobalt(II)chlorid Hexahydrat
Purity: 98.0-102.0% CAS No.: 7791-13-1 Molecular Formula: Cl2Co.6H2O Formula Weight: 237.93 (129.84anhy) Physical Form: Crystalline Density: 1.924 Appearance: Red-purple Odour: Odourless pH: 4.9 Melting Point: 87° Solubility: Soluble in water, alcohol, acetone, ether, glycerol Merck Reference: 14,2437 UN No.: 3077 MDL No.: MFCD00149652
Formel: CoCl₂·6H₂O
Molecular Weight: 237,93 g/mol
Siedepunkt: 110 °C (1013 hPa)
Schmelzpunkt: 87 °C
Dichte: 1,924
Lagertemperatur: Raumtemperatur
MDL: MFCD00149652
CAS-Nummer: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 14,2437