Nickel(II)chlorid Hexahydrat 99,30% (Metall-Basis)
Lieferant: Thermo Scientific
Gefahr
Synonyme: Nickeldichlorid Hexahydrat
12372.A3 12372.36
12372.A3EA 180 EUR
12372.A3 12372.36
Nickel(II)chlorid Hexahydrat 99,30% (Metall-Basis)
Nickel(II)chlorid Hexahydrat
Purity: 99.3% CAS No.: 7791-20-0 Molecular Formula: Cl2Ni.6H2O Formula Weight: 237.71 (129.60anhy) Physical Form: Crystalline Density: 1.92 Appearance: Green Odour: Odourless pH: 4.9 Melting Point: 140° -H2O Merck Reference: 14,6505 UN No.: 3288 MDL No.: MFCD00149809
Formel: NiCl₂·6H₂O
Molecular Weight: 237,69 g/mol
Schmelzpunkt: 140 °C (-H₂O)
Dichte: 1,92
Lagertemperatur: Raumtemperatur
MDL: MFCD00149809
CAS-Nummer: 7791-20-0
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III
Merck Index: 14,6505